Sonoco™ ThermoSafe™ Durable Insulated Upright and Chest Style Containers
Maintains product quality for your temperature-sensitive products and critical low-temperatures for laboratory-specific materials
Supplier:  Sonoco™ 858
 View more versions of this product
Catalog No. 03-001-261
Explore available promotions
Description
Description
  • Use these bulk insulated containers to effectively transport API and optimize mixed loads of refrigerated, frozen, and controlled room temperature product.
  • Keep your temperature-sensitive products at the proper temperature and preserve product quality.
  • Ideal for temporary storage in the warehouse, on the pack out line, and for the storage refrigerants and dry ice.
  • Ranging in various sizes and styles, these durable insulated containers make storing, transporting, and unloading efficient and worry-free.
Specifications
Specifications
102.9 cm
109.2 cm
121.9 cm
43 x 48 x 40.5 in. (109.2 x 121.9 x 102.9 cm)
27.0 cu. ft.
188 lb.
Interior: 37 x 41 x 29 in. (94 x 104 x 73.7 cm)
94 cm
104 cm
73.7 cm
Hard-Case Durable Insulated Transporter
40.5 in.
43 in.
48 in.
Top Closure
Transport and storage of dry ice and temperature sensitive specimens
85.3 kg
37 in.
41 in.
29 in.
High-impact Polyethylene