Sonoco™ ThermoSafe™ Durable Insulated Upright and Chest Style Containers
Maintains product quality for your temperature-sensitive products and critical low-temperatures for laboratory-specific materials
Supplier:  Sonoco™ TS860
 View more versions of this product
Catalog No. 03-001-263
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Use these bulk insulated containers to effectively transport API and optimize mixed loads of refrigerated, frozen, and controlled room temperature product.
  • Keep your temperature-sensitive products at the proper temperature and preserve product quality.
  • Ideal for temporary storage in the warehouse, on the pack out line, and for the storage refrigerants and dry ice.
  • Ranging in various sizes and styles, these durable insulated containers make storing, transporting, and unloading efficient and worry-free.
Specifications
Specifications
105.4 cm
102.9 cm
121.9 cm
43 x 48 x 41.5 in. (102.9 x 121.9 x 105.4 cm)
30 cu. ft.
243 lb.
37 in.
41 in.
31 in.
High-impact Polyethylene
41.5 in.
43 in.
48 in.
Top Closure
Transport and storage of dry ice and temperature sensitive specimens
110.2 kg
94 cm
104 cm
78.7 cm
Hard-Case Durable Insulated Transporter with Stainless Steel Inner Shell