Sonoco™ ThermoSafe™ Durable Insulated Upright and Chest Style Containers
Maintains product quality for your temperature-sensitive products and critical low-temperatures for laboratory-specific materials
Supplier:  Sonoco™ TS870
 View more versions of this product
Catalog No. 03-001-264
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Use these bulk insulated containers to effectively transport API and optimize mixed loads of refrigerated, frozen, and controlled room temperature product.
  • Keep your temperature-sensitive products at the proper temperature and preserve product quality.
  • Ideal for temporary storage in the warehouse, on the pack out line, and for the storage refrigerants and dry ice.
  • Ranging in various sizes and styles, these durable insulated containers make storing, transporting, and unloading efficient and worry-free.
Specifications
Specifications
208.3 cm
155.6 cm
123.2 cm
61.25 x 48.5 x 82 in. (155.6 x 123.2 x 208.3 cm)
74.0 cu. ft.
550 lb.
Interior: 54 x 41 x 68.5 in. (137 x 104 x 174 cm)
137 cm
104 cm
174 cm
Hard-Case Durable Insulated Transporter with Stainless Steel Inner Shell
82 in.
61.25 in.
48.5 in.
Top Closure
Transport and storage of dry ice and temperature sensitive specimens
249.5 kg
54 in.
41 in.
68.5 in.
High-impact Polyethylene