Sonoco™ ThermoSafe™ EOS
SONOCO ThermoSafe EOS is an environmentally friendly insulated shipper. ISTA 7E Thermal Performance, for 2 to 8°C seasonal packout.
Supplier:  Sonoco™ EO36KIT
 View more versions of this product
Catalog No. 03600338
Description
Description
  • Curbside recyclable product not only offers a high level of insulating value but also a soft cushioning to help protect products
  • The entire knock-down package can be placed into the recycle bin making it very easy to dispose of by the customer as ordinary recyclable trash
  • EOS is available in a range of sizes, from small to extra large
  • Note: PP24 (Catalog No. 03-531-09), is required for 2° to 8°C packout. PP24 (Catalog No. 03-531-09) polar packs, are sold separately.
Specifications
Specifications
PP24 Polar Pack Gel Packs
11.38 x 9.38 x 16.13 in. (289 x 238 x 410 mm)
2° to 8°C
Medium (5 L Payload)
Insulated Shipper
43.5 hours summer packout/72 hours winter packout
8 x 6 x 13 in. (203 x 152 x 330 mm)