Sonoco™ ThermoSafe™ EOS
SONOCO ThermoSafe EOS is an environmentally friendly insulated shipper. ISTA 7E Thermal Performance, for 2 to 8°C seasonal packout.
Supplier:  Sonoco™ EOS64KIT
 View more versions of this product
Catalog No. 03600339
Description
Description
  • Curbside recyclable product not only offers a high level of insulating value but also a soft cushioning to help protect products
  • The entire knock-down package can be placed into the recycle bin making it very easy to dispose of by the customer as ordinary recyclable trash
  • EOS is available in a range of sizes, from small to extra large
  • Note: PP24 (Catalog No. 03-531-09), is required for 2° to 8°C packout. PP24 (Catalog No. 03-531-09) polar packs, are sold separately.
Specifications
Specifications
PP24 Polar Pack Gel Packs
15.5 x 11.5 x 14.63 in. (394 x 292 x 372 mm)
2° to 8°C
Large (10 L Payload)
Insulated Shipper
43.5 hours summer packout/72 hours winter packout
12 x 8 x 11.5 in. (305 x 203 x 292 mm)