Sonoco™ ThermoSafe Insulated EPS Only foam Containers and Coolers (Small)
Large selection of cost-effective, reusable EPS foam only containers available in small, medium, and large sizes. EPS foam only shippers for the safe storage and delivery of most refrigerated or frozen products.
Supplier:  Sonoco™ TS411
 View more versions of this product
Catalog No. 03-525-4
Explore available promotions
Description
Description
  • Sizes that range 6 inches-10.5 inches in length
  • Also available in ThermoSafe Insulated EPS with Corrguate Containers and Coolers- Medium (11 inches-17 inches in length)
  • Also avaliable in ThermoSafe Insulated EPS with Corrguate Containers and Coolers- Large (19 inches-24 inches in length)
  • 200 lb. test corrugated
  • Excellent dry ice sublimation… average dry ice usage in 1-1/2 inch thick wall units is 5 pounds over 24 hours and 3-4 pounds therafter.
  • Standard 1-1/2 inch wall shippers are ideal for refrigerated shipments for 24-48 hours.
Specifications
Specifications
12 per case
11 x 9 x 10 in.
8 X 5.88 X 7 in.
Insulated Shipper
9 in.
Expanded Polystyrene Insulated