Sonoco™ ThermoSafe Insulated Storage and Transport Chests
Dry ice storage chests provide mobile, non-electrical low-temperature maintenance for a wide variety of laboratory and field applications. Ideal for dry ice or wet ice storage, hot storage, or field collection.
Supplier:  Sonoco™ TS300
 View more versions of this product
Catalog No. 03-525-2
Explore available promotions
Description
Description
Ideal for dry ice and wet ice storage, short-term frozen and hot storage.
  • Hard cased units that provide low-cost storage. Units are low maintenance and easy to clean.
  • Available in different sizes for different durations.
  • Dollies are sold separately.
  • Long term usage, all components can be replaceable.
  • Units must be filled, or dunnage added to the unit, for maximum average holding time.
Order Info
Dolly not included with chests
Specifications
Specifications
64.13 cm
71.43 cm
48.57 cm
28.12 x 19.12 x 25.25 in. (71.43 x 48.57 x 64.13 cm)
3.75 cu. ft.
40 lb.
Polyethylene
25.25 in.
28.12 in
19.12 in.
Hinged
Ideal for dry ice and wet ice storage, short-term frozen and hot storage
2 Spring Loaded Handles
Storage and Transport Chest