Sonoco™ ThermoSafe Insulated Storage and Transport Chests
Dry ice storage chests provide mobile, non-electrical low-temperature maintenance for a wide variety of laboratory and field applications. Ideal for dry ice or wet ice storage, hot storage, or field collection.
$962.47 - $3660.59
Specifications
For Use With (Application) | Ideal for dry ice and wet ice storage, short-term frozen and hot storage |
---|---|
Material | Polyethylene |
Catalog Number | Mfr. No. | Dimensions (L x W x H) | Type | Design | Closure Type | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W x H) | Type | Design | Closure Type | Price | Quantity | ||
11-676-20
|
Sonoco™
TS301 |
28.12 x 19.12 x 25.25 in. (71.43 x 48.57 x 64.13 cm) | Storage and Transport Chest | No Handles | Hinged |
Each for $3,522.32
|
|
||
11-676-11
|
Sonoco™
302 |
23 x 22 x 20.75 in. (58.42 x 55.88 x 52.70 cm) | Storage and Transport Chest | 2 Spring Loaded Handles | Hinged |
Each for $2,752.74
|
|
||
11-676-39
|
Sonoco™
TS450 |
18.25 x 16.812 x 18.75 in. (46.35 x 41 x 47.6 cm) | Storage and Transport Chest | 2 Spring Loaded Handles | Tight-fitting Plug Lid |
Each for $1,869.18
|
|
||
11-676-13
|
Sonoco™
304 |
15 x 14.062 x 13.37 in. (38.1 x 35.71 x 33.97 cm) | Storage and Transport Chest | No Handles | Tight-fitting Plug Lid |
Each for $962.47
|
|
||
03-525-2
|
Sonoco™
TS300 |
28.12 x 19.12 x 25.25 in. (71.43 x 48.57 x 64.13 cm) | Storage and Transport Chest | 2 Spring Loaded Handles | Hinged |
Each for $3,660.59
|
|
||
22-024802
|
Sonoco™
TS303 |
23 x 22 x 20.75 in. (58.42 x 55.88 x 52.70 cm) | Storage and Transport Chest | No Handles | Hinged |
Each for $2,285.68
|
|
||
03-530-27
|
Sonoco™
390 |
21.31 x 14.88 x 13.81 in. (54.12 x 37.79 x 35.07 cm) | Hard-Case Transporter | Carrying Handle | Hinged |
Each for $2,228.21
|
|
||
03-530-41
|
Sonoco™
TS422 |
15 x 14.06 x 13.38 in. (35.71 x 38.1 x 33.98 cm) | Storage and Transport Chest | Carrying Handle | Hinged |
Each for $1,513.88
|
|
||
Description
Ideal for dry ice and wet ice storage, short-term frozen and hot storage.
- Hard cased units that provide low-cost storage. Units are low maintenance and easy to clean.
- Available in different sizes for different durations.
- Dollies are sold separately.
- Long term usage, all components can be replaceable.
- Units must be filled, or dunnage added to the unit, for maximum average holding time.
Order Info
Dollies not included
Specifications
Ideal for dry ice and wet ice storage, short-term frozen and hot storage | |
Polyethylene |