missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thiazole-4-carboxaldehyde, 95%, Thermo Scientific Chemicals

Thiazole-4-carboxaldehyde, 95%, C4H3NOS, CAS Number-3364-80-5, 4-formylthiazole, 4-thiazolecarbaldehyde, 1,3-thiazole-4-carboxaldehyde, 4-formyl-1,3-thiazole, thiazole-4-carbaldehyde, thiazole-4-carboxyaldehyde, 4-thiazolecarboxaldehyde, thiazole-4-carboxaldehyde, pubchem13812, zlchem 1299, 5g

Supplier:  Thermo Scientific Chemicals H6402803

 View more versions of this product

Catalog No. AAH6402803


Explore available promotions

Description

Description

Thiazole-4-carboxaldehyde is used as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiazole-4-carboxaldehyde is used as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

Thiazole-4-carboxaldehyde
61°C to 65°C
C4H3NOS
1 g
thiazole-4-carboxaldehyde, thiazole-4-carbaldehyde, 4-thiazolecarboxaldehyde, thiazole-4-carboxyaldehyde, 4-formylthiazole, 1,3-thiazole-4-carboxaldehyde, 4-thiazolecarbaldehyde, zlchem 1299, pubchem13812, 4-formyl-1,3-thiazole
WRFKSVINLIQRKF-UHFFFAOYSA-N
1,3-thiazole-4-carbaldehyde
2763214
95%
3364-80-5
Brown
MFCD00626896
107137
Slightly soluble in water.
O=CC1=CSC=N1
113.13
113.13
Powder

Chemical Identifiers

3364-80-5
113.13
WRFKSVINLIQRKF-UHFFFAOYSA-N
2763214
O=CC1=CSC=N1
C4H3NOS
MFCD00626896
thiazole-4-carboxaldehyde, thiazole-4-carbaldehyde, 4-thiazolecarboxaldehyde, thiazole-4-carboxyaldehyde, 4-formylthiazole, 1,3-thiazole-4-carboxaldehyde, 4-thiazolecarbaldehyde, zlchem 1299, pubchem13812, 4-formyl-1,3-thiazole
1,3-thiazole-4-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c

H302-H315-H317-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only