Learn More
Thiazole-4-carboxaldehyde, 95%
Thiazole-4-carboxaldehyde, 95%, C4H3NOS, CAS Number-3364-80-5, 4-formylthiazole, 4-thiazolecarbaldehyde, 1,3-thiazole-4-carboxaldehyde, 4-formyl-1,3-thiazole, thiazole-4-carbaldehyde, thiazole-4-carboxyaldehyde, 4-thiazolecarboxaldehyde, thiazole-4-carboxaldehyde, pubchem13812, zlchem 1299, 5g | CAS: 3364-80-5 | C4H3NOS | 113.13 g/mol
$58.97 - $582.55
Chemical Identifiers
| CAS | 3364-80-5 |
|---|---|
| Molecular Formula | C4H3NOS |
| Molecular Weight (g/mol) | 113.13 |
| MDL Number | MFCD00626896 |
| InChI Key | WRFKSVINLIQRKF-UHFFFAOYSA-N |
| Synonym | thiazole-4-carboxaldehyde, thiazole-4-carbaldehyde, 4-thiazolecarboxaldehyde, thiazole-4-carboxyaldehyde, 4-formylthiazole, 1,3-thiazole-4-carboxaldehyde, 4-thiazolecarbaldehyde, zlchem 1299, pubchem13812, 4-formyl-1,3-thiazole |
| PubChem CID | 2763214 |
| IUPAC Name | 1,3-thiazole-4-carbaldehyde |
| SMILES | O=CC1=CSC=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH64028MD
|
Thermo Scientific Chemicals
H64028MD |
250 mg |
N/A
|
|
|||||
|
AAH6402803
|
Thermo Scientific Chemicals
H6402803 |
1 g |
N/A
|
|
|||||
|
AAH6402806
|
Thermo Scientific Chemicals
H6402806 |
5 g |
N/A
|
|
|||||
Description
Thiazole-4-carboxaldehyde is used as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThiazole-4-carboxaldehyde is used as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
| 3364-80-5 | |
| 113.13 | |
| WRFKSVINLIQRKF-UHFFFAOYSA-N | |
| 2763214 | |
| O=CC1=CSC=N1 |
| C4H3NOS | |
| MFCD00626896 | |
| thiazole-4-carboxaldehyde, thiazole-4-carbaldehyde, 4-thiazolecarboxaldehyde, thiazole-4-carboxyaldehyde, 4-formylthiazole, 1,3-thiazole-4-carboxaldehyde, 4-thiazolecarbaldehyde, zlchem 1299, pubchem13812, 4-formyl-1,3-thiazole | |
| 1,3-thiazole-4-carbaldehyde |
Specifications
| 3364-80-5 | |
| Brown | |
| MFCD00626896 | |
| 107137 | |
| Slightly soluble in water. | |
| O=CC1=CSC=N1 | |
| 113.13 | |
| 113.13 | |
| Powder |
| 61°C to 65°C | |
| C4H3NOS | |
| 250 mg | |
| thiazole-4-carboxaldehyde, thiazole-4-carbaldehyde, 4-thiazolecarboxaldehyde, thiazole-4-carboxyaldehyde, 4-formylthiazole, 1,3-thiazole-4-carboxaldehyde, 4-thiazolecarbaldehyde, zlchem 1299, pubchem13812, 4-formyl-1,3-thiazole | |
| WRFKSVINLIQRKF-UHFFFAOYSA-N | |
| 1,3-thiazole-4-carbaldehyde | |
| 2763214 | |
| 95% | |
| Thiazole-4-carboxaldehyde |
Safety and Handling
GHS H Statement
H301-H315-H319-H317-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
H302-H315-H317-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only