Greiner Bio-One ThinCert™ 96 Well HTS Insert
Thincert™ 96 Well HTS inserts are indispensable tools for high-throughput screening and assays. Designed for precision, they enable researchers to elevate the quality and efficiency of their work.
Supplier:  Greiner Bio-One 655169
 View more versions of this product
Catalog No. 01-670-252
Explore available promotions
Description
Description
  • Optimal tool for generating tissue models (e.g., endothelia and epithelia) for transport, uptake, or co-culture studies in submersed as well as air-lift culture with an automation-friendly format
  • By combining high membrane permeability and best possible transparency, Greiner Bio-One is offering a unique 96 well insert version, exclusively designed for applications that require both high diffusion rates as well as microscopic readouts.
  • Special pore arrangement and a high pore density of 2 x 107/cm2.
  • Patented assembly technology: robust and reliable bonding of membrane 
  • Excellent membrane flatness: optimal and reproducible cell culture conditions 
  • Optimized fit: prevention of media wicking
  • Enlarged access ports: convenient access for pipettes, robots and TEER electrodes
Specifications
Specifications
No Barcode
8 x 12
Receiver Plate
1.25 to 3.12 μL
120 to 300 μL
Non-treated
Standard
Clear
Polycarbonate
96
Sterile
Flat Bottom (F-well)
Product Suggestions
Product Suggestions