Greiner Bio-One ThinCert™ 96 Well HTS Insert
Thincert™ 96 Well HTS inserts are indispensable tools for high-throughput screening and assays. Designed for precision, they enable researchers to elevate the quality and efficiency of their work.
$2169.71 - $8737.76
Specifications
Barcode No Barcode
Format Standard
Array 8 x 12
Color Clear
Material Polycarbonate
View More Specs
Products 4
Catalog Number Mfr. No. Product Type Surface Treatment Price Quantity  
Catalog Number Mfr. No. Product Type Surface Treatment Price Quantity  
01-670-252
View Documents
Greiner Bio-One
655169
Receiver Plate Non-treated
Case of 32 Each for $2,169.71
01-670-254
View Documents
Greiner Bio-One
655167
Receiver Plate Tissue Culture
Case of 32 Each for $2,314.00
01-670-253
View Documents
Greiner Bio-One
655640
Insert with Permeable Membrane Tissue Culture
Case of 5 Each for $8,385.15
01-670-255
View Documents
Greiner Bio-One
655641
Insert with Transparent Membrane Tissue Culture
Case of 5 Each for $8,737.76
Description
Description
  • Optimal tool for generating tissue models (e.g., endothelia and epithelia) for transport, uptake, or co-culture studies in submersed as well as air-lift culture with an automation-friendly format
  • By combining high membrane permeability and best possible transparency, Greiner Bio-One is offering a unique 96 well insert version, exclusively designed for applications that require both high diffusion rates as well as microscopic readouts.
  • Special pore arrangement and a high pore density of 2 x 107/cm2.
  • Patented assembly technology: robust and reliable bonding of membrane 
  • Excellent membrane flatness: optimal and reproducible cell culture conditions 
  • Optimized fit: prevention of media wicking
  • Enlarged access ports: convenient access for pipettes, robots and TEER electrodes
Specifications
Specifications
No Barcode
8 x 12
Polycarbonate
Sterile
Standard
Clear
96
Flat Bottom (F-well)
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'