missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Greiner Bio-One ThinCert™ 96 Well HTS Insert
Thincert™ 96 Well HTS inserts are indispensable tools for high-throughput screening and assays. Designed for precision, they enable researchers to elevate the quality and efficiency of their work.
Supplier: Greiner Bio-One 655640
Description
- Optimal tool for generating tissue models (e.g., endothelia and epithelia) for transport, uptake, or co-culture studies in submersed as well as air-lift culture with an automation-friendly format
- By combining high membrane permeability and best possible transparency, Greiner Bio-One is offering a unique 96 well insert version, exclusively designed for applications that require both high diffusion rates as well as microscopic readouts.
- Special pore arrangement and a high pore density of 2 x 107/cm2.
- Patented assembly technology: robust and reliable bonding of membrane
- Excellent membrane flatness: optimal and reproducible cell culture conditions
- Optimized fit: prevention of media wicking
- Enlarged access ports: convenient access for pipettes, robots and TEER electrodes
Specifications
No Barcode | |
8 x 12 | |
Insert with Permeable Membrane | |
0.15 to 1.7 μL | |
15 to 160 μL | |
Tissue Culture |
Standard | |
Clear | |
Polycarbonate | |
96 | |
Sterile | |
Flat Bottom (F-well) |