missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 424530250

 View more versions of this product

Catalog No. AC424530250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Thioacetamide
112.0°C to 114.0°C
1.3700g/mL
Authentic
Glass bottle
CH3CSNH2
25 g
04,502
15, 9472
Solubility in water: 163g/L (25°C). Other solubilities: 26.4g/100g ethanol, sparingly soluble in ether, soluble in benzene
CC(N)=S
75.13
CHEBI:32497
99+%
ACS Reagent
62-55-5
White to Beige
0.05% max.
99% min. (Argentometry)
C2H5NS
MFCD00008070
02, 232
1.37
thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide
YUKQRDCYNOVPGJ-UHFFFAOYSA-N
ethanethioamide
2723949
75.13
(2% soln.) Passes test
Crystalline Powder

Chemical Identifiers

62-55-5
75.13
YUKQRDCYNOVPGJ-UHFFFAOYSA-N
2723949
ethanethioamide
C2H5NS
MFCD00008070
thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide
CHEBI:32497
CC(N)=S
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause cancer.
Harmful to aquatic life with long lasting effects.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water

GHS Signal Word: Danger

EINECSNumber : 200-541-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only