Learn More
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals
$132.47 - $1474.30
Chemical Identifiers
CAS | 62-55-5 |
---|---|
Molecular Formula | C2H5NS |
Molecular Weight (g/mol) | 75.13 |
MDL Number | MFCD00008070 |
InChI Key | YUKQRDCYNOVPGJ-UHFFFAOYSA-N |
Synonym | thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide |
PubChem CID | 2723949 |
ChEBI | CHEBI:32497 |
IUPAC Name | ethanethioamide |
SMILES | CC(N)=S |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC424530250
|
Thermo Scientific Chemicals
424530250 |
25 g | Glass bottle |
Each for $132.47
|
|
||||
AC424531000
|
Thermo Scientific Chemicals
424531000 |
100 g | Glass bottle |
Each for $396.53
Case of 6 Each for $0.00
|
|
||||
AC424535000
|
Thermo Scientific Chemicals
424535000 |
500 g | Glass bottle |
Each for $1,474.30
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62-55-5 | |
75.13 | |
YUKQRDCYNOVPGJ-UHFFFAOYSA-N | |
2723949 | |
ethanethioamide |
C2H5NS | |
MFCD00008070 | |
thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide | |
CHEBI:32497 | |
CC(N)=S |
Specifications
62-55-5 | |
White to Beige | |
0.05% max. | |
99% min. (Argentometry) | |
C2H5NS | |
MFCD00008070 | |
02, 232 | |
1.37 | |
thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide | |
YUKQRDCYNOVPGJ-UHFFFAOYSA-N | |
ethanethioamide | |
2723949 | |
75.13 | |
(2% soln.) Passes test | |
Crystalline Powder |
112.0°C to 114.0°C | |
1.3700g/mL | |
Authentic | |
Glass bottle | |
CH3CSNH2 | |
25 g | |
04,502 | |
15, 9472 | |
Solubility in water: 163g/L (25°C). Other solubilities: 26.4g/100g ethanol, sparingly soluble in ether, soluble in benzene | |
CC(N)=S | |
75.13 | |
CHEBI:32497 | |
99+% | |
ACS Reagent | |
Thioacetamide |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause cancer.
Harmful to aquatic life with long lasting effects.
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water
GHS Signal Word: Danger
EINECSNumber : 200-541-4
RUO – Research Use Only