missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thioacetic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0330522

 View more versions of this product

Catalog No. AAL0330522


Explore available promotions

Description

Description

Thioacetic acid is a reagent for introduction of the thiol group into organic molecules. It is used as flavor & Fragrance Intermediates. It is used to manufacture captopril(antihypertension) and spironolac-tone (diuretic).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thioacetic acid is a reagent for introduction of the thiol group into organic molecules. It is used as flavor & Fragrance Intermediates. It is used to manufacture captopril(antihypertension) and spironolac-tone (diuretic).

Solubility
Soluble in water. (27g/L) at 16°C.

Notes
Protect from heat. Store away from oxidizing agents. Incompatible with strong bases, strong oxidizing agents, metals.
Specifications

Specifications

Thioacetic acid
-17°C
97°C
Stench
1.464
100 g
773684
thioacetic acid, ethanethioic acid, thiacetic acid, thioacetic s-acid, acetyl mercaptan, ethanethiolic acid, thiolacetic acid, thionoacetic acid, methanecarbothiolic acid, ch3cosh
DUYAAUVXQSMXQP-UHFFFAOYSA-N
ethanethioic S-acid
10484
76.12
507-09-5
1.068
18°C (64°F)
C2H4OS
MFCD00004853
UN2436
14,9320
Soluble in water. (27g/L) at 16°C.
CC(S)=O
76.11
CHEBI:16555
97%

Chemical Identifiers

507-09-5
76.11
DUYAAUVXQSMXQP-UHFFFAOYSA-N
10484
ethanethioic S-acid
C2H4OS
MFCD00004853
thioacetic acid, ethanethioic acid, thiacetic acid, thioacetic s-acid, acetyl mercaptan, ethanethiolic acid, thiolacetic acid, thionoacetic acid, methanecarbothiolic acid, ch3cosh
CHEBI:16555
CC(S)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318-H302-H332-H317
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

P210-P233-P240-P241-P242-P243-P261-P264b-P270-P272-P280-P301+P310-P303+P361+P353-P305+P351+P338-P310-P330-P333+P313-P363-P370+P378q-P501c

H225-H301-H317-H318-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: THIOACETIC ACID

EINECSNumber : 208-063-8

RTECSNumber : AJ5600000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only