missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thioacetic acid, 97%, Thermo Scientific Chemicals

$61.73 - $272.10

Chemical Identifiers

CAS 507-09-5
Molecular Formula C2H4OS
Molecular Weight (g/mol) 76.11
MDL Number MFCD00004853
InChI Key DUYAAUVXQSMXQP-UHFFFAOYSA-N
Synonym thioacetic acid, ethanethioic acid, thiacetic acid, thioacetic s-acid, acetyl mercaptan, ethanethiolic acid, thiolacetic acid, thionoacetic acid, methanecarbothiolic acid, ch3cosh
PubChem CID 10484
ChEBI CHEBI:16555
IUPAC Name ethanethioic S-acid
SMILES CC(S)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0330514
View Documents
Thermo Scientific Chemicals
L0330514
25 g
Each for $61.73
 
AAL0330522
View Documents
Thermo Scientific Chemicals
L0330522
100 g
Each for $92.96
 
AAL0330536
View Documents
Thermo Scientific Chemicals
L0330536
500 g
Each for $272.10
 
Description

Description

Thioacetic acid is a reagent for introduction of the thiol group into organic molecules. It is used as flavor & Fragrance Intermediates. It is used to manufacture captopril(antihypertension) and spironolac-tone (diuretic).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thioacetic acid is a reagent for introduction of the thiol group into organic molecules. It is used as flavor & Fragrance Intermediates. It is used to manufacture captopril(antihypertension) and spironolac-tone (diuretic).

Solubility
Soluble in water. (27g/L) at 16°C.

Notes
Protect from heat. Store away from oxidizing agents. Incompatible with strong bases, strong oxidizing agents, metals.
Specifications

Chemical Identifiers

507-09-5
76.11
DUYAAUVXQSMXQP-UHFFFAOYSA-N
10484
ethanethioic S-acid
C2H4OS
MFCD00004853
thioacetic acid, ethanethioic acid, thiacetic acid, thioacetic s-acid, acetyl mercaptan, ethanethiolic acid, thiolacetic acid, thionoacetic acid, methanecarbothiolic acid, ch3cosh
CHEBI:16555
CC(S)=O

Specifications

507-09-5
1.068
18°C (64°F)
C2H4OS
MFCD00004853
UN2436
14,9320
Soluble in water. (27g/L) at 16°C.
CC(S)=O
76.11
CHEBI:16555
97%
-17°C
97°C
Stench
1.464
25 g
773684
thioacetic acid, ethanethioic acid, thiacetic acid, thioacetic s-acid, acetyl mercaptan, ethanethiolic acid, thiolacetic acid, thionoacetic acid, methanecarbothiolic acid, ch3cosh
DUYAAUVXQSMXQP-UHFFFAOYSA-N
ethanethioic S-acid
10484
76.12
Thioacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318-H302-H332-H317
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

P210-P233-P240-P241-P242-P243-P261-P264b-P270-P272-P280-P301+P310-P303+P361+P353-P305+P351+P338-P310-P330-P333+P313-P363-P370+P378q-P501c

H225-H301-H317-H318-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: THIOACETIC ACID

EINECSNumber : 208-063-8

RTECSNumber : AJ5600000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only