missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thiochroman-4-one, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 153480100

 View more versions of this product

Catalog No. AC153480100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Thiochroman-4-one
154°C (12.0mmHg)
Authentic
Glass bottle
1.6395
10g
thiochroman-4-one, 4h-1-benzothiopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-thiochromen-4-one, 3,4-dihydro-2h-1-benzothiopyran-4-one, 4h-1-benzothiopyran-4-one,3-dihydro, 2,3-dihydro-1-benzothiopyran-4-one, 4-thiochromanone, thiochroman-4-on, 1-thiochroman-4-one
C1CSC2=CC=CC=C2C1=O
164.22
164.22
3528-17-4
>110°C
97.5% min. (GC)
C9H8OS
MFCD00006882
17, II, 336
CVQSWZMJOGOPAV-UHFFFAOYSA-N
2,3-dihydrothiochromen-4-one
19048
98%

Chemical Identifiers

3528-17-4
164.22
CVQSWZMJOGOPAV-UHFFFAOYSA-N
19048
C1CSC2=CC=CC=C2C1=O
C9H8OS
MFCD00006882
thiochroman-4-one, 4h-1-benzothiopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-thiochromen-4-one, 3,4-dihydro-2h-1-benzothiopyran-4-one, 4h-1-benzothiopyran-4-one,3-dihydro, 2,3-dihydro-1-benzothiopyran-4-one, 4-thiochromanone, thiochroman-4-on, 1-thiochroman-4-one
2,3-dihydrothiochromen-4-one
Safety and Handling

Safety and Handling

EINECSNumber : 222-548-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only