missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thiochroman-4-one, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 3528-17-4 |
---|---|
Molecular Formula | C9H8OS |
Molecular Weight (g/mol) | 164.22 |
MDL Number | MFCD00006882 |
InChI Key | CVQSWZMJOGOPAV-UHFFFAOYSA-N |
Synonym | thiochroman-4-one, 4h-1-benzothiopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-thiochromen-4-one, 3,4-dihydro-2h-1-benzothiopyran-4-one, 4h-1-benzothiopyran-4-one,3-dihydro, 2,3-dihydro-1-benzothiopyran-4-one, 4-thiochromanone, thiochroman-4-on, 1-thiochroman-4-one |
PubChem CID | 19048 |
IUPAC Name | 2,3-dihydrothiochromen-4-one |
SMILES | C1CSC2=CC=CC=C2C1=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC153480050
|
Thermo Scientific Chemicals
153480050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC153480100
|
Thermo Scientific Chemicals
153480100 |
10g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
3528-17-4 | |
164.22 | |
CVQSWZMJOGOPAV-UHFFFAOYSA-N | |
19048 | |
C1CSC2=CC=CC=C2C1=O |
C9H8OS | |
MFCD00006882 | |
thiochroman-4-one, 4h-1-benzothiopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-thiochromen-4-one, 3,4-dihydro-2h-1-benzothiopyran-4-one, 4h-1-benzothiopyran-4-one,3-dihydro, 2,3-dihydro-1-benzothiopyran-4-one, 4-thiochromanone, thiochroman-4-on, 1-thiochroman-4-one | |
2,3-dihydrothiochromen-4-one |
Specifications
3528-17-4 | |
>110°C | |
97.5% min. (GC) | |
C9H8OS | |
MFCD00006882 | |
17, II, 336 | |
CVQSWZMJOGOPAV-UHFFFAOYSA-N | |
2,3-dihydrothiochromen-4-one | |
19048 | |
98% |
154°C (12.0mmHg) | |
Authentic | |
Glass bottle | |
1.6395 | |
5g | |
thiochroman-4-one, 4h-1-benzothiopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-thiochromen-4-one, 3,4-dihydro-2h-1-benzothiopyran-4-one, 4h-1-benzothiopyran-4-one,3-dihydro, 2,3-dihydro-1-benzothiopyran-4-one, 4-thiochromanone, thiochroman-4-on, 1-thiochroman-4-one | |
C1CSC2=CC=CC=C2C1=O | |
164.22 | |
164.22 | |
Thiochroman-4-one |
Safety and Handling
EINECSNumber : 222-548-1
RUO â Research Use Only