missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thioglycolic acid, 99.94%, Bacteriological grade, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210293390
Specifications
Thioglycolic acid | |
White | |
C2H3NaO2S | |
sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate | |
C(C(=O)[O-])S.[Na+] | |
114.094 | |
CHEBI:86481 | |
0.9994 | |
Crystalline Powder |
367-51-1 | |
∽1.32g/mL | |
500 g | |
GNBVPFITFYNRCN-UHFFFAOYSA-M | |
sodium;2-sulfanylacetate | |
23690444 | |
114.11 | |
Bacteriological |
Chemical Identifiers
367-51-1 | |
114.094 | |
sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate | |
CHEBI:86481 | |
C(C(=O)[O-])S.[Na+] |
C2H3NaO2S | |
GNBVPFITFYNRCN-UHFFFAOYSA-M | |
23690444 | |
sodium;2-sulfanylacetate |