missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thioglycolic acid, 99.94%, Bacteriological grade, MP Biomedicals™
$168.10 - $569.08
Chemical Identifiers
CAS | 367-51-1 |
---|---|
Molecular Formula | C2H3NaO2S |
Molecular Weight (g/mol) | 114.094 |
InChI Key | GNBVPFITFYNRCN-UHFFFAOYSA-M |
Synonym | sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate |
PubChem CID | 23690444 |
ChEBI | CHEBI:86481 |
IUPAC Name | sodium;2-sulfanylacetate |
SMILES | C(C(=O)[O-])S.[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10293380
|
MP Biomedicals Inc
0210293380 |
100 g |
Each for $168.10
|
|
|||||
ICN10293390
|
MP Biomedicals Inc
0210293390 |
500 g |
Each for $569.08
|
|
|||||
Chemical Identifiers
367-51-1 | |
114.094 | |
sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate | |
CHEBI:86481 | |
C(C(=O)[O-])S.[Na+] |
C2H3NaO2S | |
GNBVPFITFYNRCN-UHFFFAOYSA-M | |
23690444 | |
sodium;2-sulfanylacetate |
Specifications
367-51-1 | |
C2H3NaO2S | |
sodium thioglycolate, sodium mercaptoacetate, mercaptoacetic acid sodium salt, mollescal sf, sodium 2-sulfanylacetate, thioglycolate sodium, sodium thioglycollate, mercaptoacetic acid, sodium salt, erhavit d, sodium 2-mercatoethanoate | |
C(C(=O)[O-])S.[Na+] | |
114.094 | |
CHEBI:86481 | |
0.9994 | |
Crystalline Powder |
White | |
100 g | |
GNBVPFITFYNRCN-UHFFFAOYSA-M | |
sodium;2-sulfanylacetate | |
23690444 | |
114.11 | |
Bacteriological | |
Thioglycolic acid |