missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thiophene-2,5-dicarboxylic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1029306

 View more versions of this product

Catalog No. AAL1029306


Explore available promotions

Description

Description

Thiophene-2,5-dicarboxylic acid may be used in the synthesis new cobalt(II) thiophenedicarboxylate coordination polymer and novel mesogenic thiophene based supramolecular liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2,5-dicarboxylic acid may be used in the synthesis new cobalt(II) thiophenedicarboxylate coordination polymer and novel mesogenic thiophene based supramolecular liquid crystals.

Solubility
Very soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Specifications

Thiophene-2,5-dicarboxylic acid
355°C to 360°C
MFCD00016896
136781
Very soluble in water.
C1=C(SC(=C1)C(=O)O)C(=O)O
172.154
172.16
4282-31-9
C6H4O4S
5 g
2,5-thiophenedicarboxylic acid, 2,5-dicarboxythiophene, unii-5q8a2g9y8a, 2,5-thiophene dicarboxylic acid, pubchem5580, maybridge4_002021, acmc-1aj5p, ksc182i5d, 2,5-thiophene-dicarboxylic acid, thiophene-2.5-dicarboxylic acid
YCGAZNXXGKTASZ-UHFFFAOYSA-N
thiophene-2,5-dicarboxylic acid
20273
97%

Chemical Identifiers

4282-31-9
172.154
YCGAZNXXGKTASZ-UHFFFAOYSA-N
20273
C1=C(SC(=C1)C(=O)O)C(=O)O
C6H4O4S
MFCD00016896
2,5-thiophenedicarboxylic acid, 2,5-dicarboxythiophene, unii-5q8a2g9y8a, 2,5-thiophene dicarboxylic acid, pubchem5580, maybridge4_002021, acmc-1aj5p, ksc182i5d, 2,5-thiophene-dicarboxylic acid, thiophene-2.5-dicarboxylic acid
thiophene-2,5-dicarboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 224-284-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only