missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2,5-dicarboxylic acid, 97%

CAS: 4282-31-9 | C6H4O4S | 172.154 g/mol

$47.14 - $346.00

Chemical Identifiers

CAS 4282-31-9
Molecular Formula C6H4O4S
Molecular Weight (g/mol) 172.154
MDL Number MFCD00016896
InChI Key YCGAZNXXGKTASZ-UHFFFAOYSA-N
Synonym 2,5-thiophenedicarboxylic acid, 2,5-dicarboxythiophene, unii-5q8a2g9y8a, 2,5-thiophene dicarboxylic acid, pubchem5580, maybridge4_002021, acmc-1aj5p, ksc182i5d, 2,5-thiophene-dicarboxylic acid, thiophene-2.5-dicarboxylic acid
PubChem CID 20273
IUPAC Name thiophene-2,5-dicarboxylic acid
SMILES C1=C(SC(=C1)C(=O)O)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1029303
View Documents
Thermo Scientific Chemicals
L1029303
1 g N/A
Only null left
 
AAL1029306
View Documents
Thermo Scientific Chemicals
L1029306
5 g N/A
Only null left
 
AAL1029314
View Documents
Thermo Scientific Chemicals
L1029314
25 g N/A
Only null left
 
Description

Description

Thiophene-2,5-dicarboxylic acid may be used in the synthesis new cobalt(II) thiophenedicarboxylate coordination polymer and novel mesogenic thiophene based supramolecular liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2,5-dicarboxylic acid may be used in the synthesis new cobalt(II) thiophenedicarboxylate coordination polymer and novel mesogenic thiophene based supramolecular liquid crystals.

Solubility
Very soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Chemical Identifiers

4282-31-9
172.154
YCGAZNXXGKTASZ-UHFFFAOYSA-N
20273
C1=C(SC(=C1)C(=O)O)C(=O)O
C6H4O4S
MFCD00016896
2,5-thiophenedicarboxylic acid, 2,5-dicarboxythiophene, unii-5q8a2g9y8a, 2,5-thiophene dicarboxylic acid, pubchem5580, maybridge4_002021, acmc-1aj5p, ksc182i5d, 2,5-thiophene-dicarboxylic acid, thiophene-2.5-dicarboxylic acid
thiophene-2,5-dicarboxylic acid

Specifications

4282-31-9
C6H4O4S
1 g
2,5-thiophenedicarboxylic acid, 2,5-dicarboxythiophene, unii-5q8a2g9y8a, 2,5-thiophene dicarboxylic acid, pubchem5580, maybridge4_002021, acmc-1aj5p, ksc182i5d, 2,5-thiophene-dicarboxylic acid, thiophene-2.5-dicarboxylic acid
YCGAZNXXGKTASZ-UHFFFAOYSA-N
thiophene-2,5-dicarboxylic acid
20273
97%
355°C to 360°C
MFCD00016896
136781
Very soluble in water.
C1=C(SC(=C1)C(=O)O)C(=O)O
172.154
172.16
Thiophene-2,5-dicarboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 224-284-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only