missing translation for 'onlineSavingsMsg'
Learn More

Thiophene, 99.5%, extra pure, benzene free

CAS: 110-02-1 | C4H4S | 84.14 g/mol

Supplier:  Thermo Scientific Chemicals 342595000

 View more versions of this product

Catalog No. AC342595000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Thiophene
110-02-1
1.0650g/mL
−9°C
99.4% min. (GC)
C4H4S
MFCD00005413
17, 29
15, 9506
Solubility in water: insoluble. Other solubilities: miscible with most organic solvents
S1C=CC=C1
84.14
CHEBI:30856
99.5%
Liquid
99.5%
-38.0°C
84.0°C
Authentic
Glass bottle
1.5270 to 1.5290
500 mL
1.065
thiofuran, thiole, thiophen, thiotetrole, thiacyclopentadiene, divinylene sulfide, thiaphene, thiofuram, thiofurfuran, furan, thio
YTPLMLYBLZKORZ-UHFFFAOYSA-N
thiophene
8030
84.14
Extra Pure

Chemical Identifiers

110-02-1
84.14
YTPLMLYBLZKORZ-UHFFFAOYSA-N
8030
thiophene
C4H4S
MFCD00005413
thiofuran, thiole, thiophen, thiotetrole, thiacyclopentadiene, divinylene sulfide, thiaphene, thiofuram, thiofurfuran, furan, thio
CHEBI:30856
S1C=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapour.
Harmful if swallowed.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi

GHS Signal Word: Danger

EINECSNumber : 203-729-4

RTECSNumber : XM7350000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only