Learn More
Thiophene, 99.5%, extra pure, benzene free, Thermo Scientific Chemicals
$62.43 - $263.84
Chemical Identifiers
CAS | 110-02-1 |
---|---|
Molecular Formula | C4H4S |
Molecular Weight (g/mol) | 84.14 |
MDL Number | MFCD00005413 |
InChI Key | YTPLMLYBLZKORZ-UHFFFAOYSA-N |
Synonym | thiofuran, thiole, thiophen, thiotetrole, thiacyclopentadiene, divinylene sulfide, thiaphene, thiofuram, thiofurfuran, furan, thio |
PubChem CID | 8030 |
ChEBI | CHEBI:30856 |
IUPAC Name | thiophene |
SMILES | S1C=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC342591000
|
Thermo Scientific Chemicals
342591000 |
100 mL | Glass bottle |
N/A
|
|
||||
AC342595000
|
Thermo Scientific Chemicals
342595000 |
500 mL | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-02-1 | |
84.14 | |
YTPLMLYBLZKORZ-UHFFFAOYSA-N | |
8030 | |
thiophene |
C4H4S | |
MFCD00005413 | |
thiofuran, thiole, thiophen, thiotetrole, thiacyclopentadiene, divinylene sulfide, thiaphene, thiofuram, thiofurfuran, furan, thio | |
CHEBI:30856 | |
S1C=CC=C1 |
Specifications
110-02-1 | |
1.0650g/mL | |
−9°C | |
99.4% min. (GC) | |
C4H4S | |
MFCD00005413 | |
17, 29 | |
15, 9506 | |
Solubility in water: insoluble. Other solubilities: miscible with most organic solvents | |
S1C=CC=C1 | |
84.14 | |
CHEBI:30856 | |
99.5% | |
Liquid |
-38.0°C | |
84.0°C | |
Authentic | |
Glass bottle | |
1.5270 to 1.5290 | |
100 mL | |
1.065 | |
thiofuran, thiole, thiophen, thiotetrole, thiacyclopentadiene, divinylene sulfide, thiaphene, thiofuram, thiofurfuran, furan, thio | |
YTPLMLYBLZKORZ-UHFFFAOYSA-N | |
thiophene | |
8030 | |
84.14 | |
Extra Pure | |
Thiophene, 99.5% |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
Harmful if swallowed.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
GHS Signal Word: Danger
EINECSNumber : 203-729-4
RTECSNumber : XM7350000
TSCA : TSCA
RUO – Research Use Only