missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thiosalicylic Acid 90.0+%, TCI America™
Supplier: TCI America M005425G
Specifications
Thiosalicylic Acid | |
147-93-3 | |
MFCD00004836 | |
NBOMNTLFRHMDEZ-UHFFFAOYSA-N | |
2-sulfanylbenzoic acid | |
5443 | |
154.18 | |
25 g |
167°C | |
C7H6O2S | |
thiosalicylic acid, 2-mercaptobenzoic acid, o-mercaptobenzoic acid, 2-thiosalicylic acid, o-thiosalicylic acid, 2-carboxythiophenol, o-benzoic acid thiol, o-carboxythiophenol, o-sulfhydrylbenzoic acid, benzoic acid, 2-mercapto | |
C1=CC=C(C(=C1)C(=O)O)S | |
154.183 | |
CHEBI:59124 | |
≥90.0% (T) | |
Crystalline Powder |
Chemical Identifiers
147-93-3 | |
154.183 | |
NBOMNTLFRHMDEZ-UHFFFAOYSA-N | |
5443 | |
2-sulfanylbenzoic acid |
C7H6O2S | |
MFCD00004836 | |
thiosalicylic acid, 2-mercaptobenzoic acid, o-mercaptobenzoic acid, 2-thiosalicylic acid, o-thiosalicylic acid, 2-carboxythiophenol, o-benzoic acid thiol, o-carboxythiophenol, o-sulfhydrylbenzoic acid, benzoic acid, 2-mercapto | |
CHEBI:59124 | |
C1=CC=C(C(=C1)C(=O)O)S |
Safety and Handling
EINECSNumber : (3)-1657
RTECSNumber : DH3325000
TSCA : Yes