missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thiosalicylic Acid 90.0+%, TCI America™

$43.25 - $500.00

Chemical Identifiers

CAS 147-93-3
Molecular Formula C7H6O2S
Molecular Weight (g/mol) 154.183
MDL Number MFCD00004836
InChI Key NBOMNTLFRHMDEZ-UHFFFAOYSA-N
Synonym thiosalicylic acid, 2-mercaptobenzoic acid, o-mercaptobenzoic acid, 2-thiosalicylic acid, o-thiosalicylic acid, 2-carboxythiophenol, o-benzoic acid thiol, o-carboxythiophenol, o-sulfhydrylbenzoic acid, benzoic acid, 2-mercapto
PubChem CID 5443
ChEBI CHEBI:59124
IUPAC Name 2-sulfanylbenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M005425G
View Documents
TCI America
M005425G
25 g
Each for $43.25
 
M0054500G
View Documents
TCI America
M0054500G
500 g
Each for $500.00
 
Description

Description

Specifications

Chemical Identifiers

147-93-3
154.183
NBOMNTLFRHMDEZ-UHFFFAOYSA-N
5443
2-sulfanylbenzoic acid
C7H6O2S
MFCD00004836
thiosalicylic acid, 2-mercaptobenzoic acid, o-mercaptobenzoic acid, 2-thiosalicylic acid, o-thiosalicylic acid, 2-carboxythiophenol, o-benzoic acid thiol, o-carboxythiophenol, o-sulfhydrylbenzoic acid, benzoic acid, 2-mercapto
CHEBI:59124
C1=CC=C(C(=C1)C(=O)O)S

Specifications

167°C
C7H6O2S
thiosalicylic acid, 2-mercaptobenzoic acid, o-mercaptobenzoic acid, 2-thiosalicylic acid, o-thiosalicylic acid, 2-carboxythiophenol, o-benzoic acid thiol, o-carboxythiophenol, o-sulfhydrylbenzoic acid, benzoic acid, 2-mercapto
C1=CC=C(C(=C1)C(=O)O)S
154.183
CHEBI:59124
≥90.0% (T)
Crystalline Powder
147-93-3
MFCD00004836
NBOMNTLFRHMDEZ-UHFFFAOYSA-N
2-sulfanylbenzoic acid
5443
154.18
25 g
Thiosalicylic Acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1657

RTECSNumber : DH3325000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'