missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thiosalicylic Acid 90.0+%, TCI America™

Supplier:  TCI America M0054500G

 View more versions of this product

Catalog No. M0054500G


Explore available promotions

Description

Description

Specifications

Specifications

Thiosalicylic Acid
147-93-3
MFCD00004836
NBOMNTLFRHMDEZ-UHFFFAOYSA-N
2-sulfanylbenzoic acid
5443
154.18
500 g
167°C
C7H6O2S
thiosalicylic acid, 2-mercaptobenzoic acid, o-mercaptobenzoic acid, 2-thiosalicylic acid, o-thiosalicylic acid, 2-carboxythiophenol, o-benzoic acid thiol, o-carboxythiophenol, o-sulfhydrylbenzoic acid, benzoic acid, 2-mercapto
C1=CC=C(C(=C1)C(=O)O)S
154.183
CHEBI:59124
≥90.0% (T)
Crystalline Powder

Chemical Identifiers

147-93-3
154.183
NBOMNTLFRHMDEZ-UHFFFAOYSA-N
5443
2-sulfanylbenzoic acid
C7H6O2S
MFCD00004836
thiosalicylic acid, 2-mercaptobenzoic acid, o-mercaptobenzoic acid, 2-thiosalicylic acid, o-thiosalicylic acid, 2-carboxythiophenol, o-benzoic acid thiol, o-carboxythiophenol, o-sulfhydrylbenzoic acid, benzoic acid, 2-mercapto
CHEBI:59124
C1=CC=C(C(=C1)C(=O)O)S
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1657

RTECSNumber : DH3325000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions