missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thiourea, 99%, extra pure, Thermo Scientific Chemicals

Thiourea, Thiocarbamide, CAS # 62-56-6, is an analogue of urea with a single sulfur-substituted oxygen.

Supplier:  Thermo Scientific Chemicals 138915000

 View more versions of this product

Catalog No. AC138915000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Thiourea is a derivative of the biological compound, urea
• Thiourea is highly toxic when it decomposes and acts as a carcinogen

Applications

• Thiourea can be used as a denaturant in lysis buffers and similar sample buffers that require solubilization of proteins
• Thiourea can act as an antioxidant and chromophore
• Thiourea can be used in the study of cellular redox, autophagy, and aging

Specifications

Specifications

Thiourea
171.0°C to 175.0°C
0.5% max.
98.5% min. (Iodimetry)
CH4N2S
MFCD00008067
03, 180
15, 9521
Solubility in water: 136g/L (20°C). Other solubilities: 12g/100g methanol (20°C),soluble in isopropylalcohol,butanol ,pentanol,sparingly soluble in ether
NC(N)=S
76.12
CHEBI:36946
99%
Crystalline Powder or Crystals
62-56-6
White
Authentic
Plastic bottle
H2NCSNH2
500 g
01,1164; 02,412; 03,290; 06,586; 11,519; 15,308
thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio
UMGDCJDMYOKAJW-UHFFFAOYSA-N
thiourea
2723790
76.12
Extra Pure

Chemical Identifiers

62-56-6
76.12
UMGDCJDMYOKAJW-UHFFFAOYSA-N
2723790
thiourea
CH4N2S
MFCD00008067
thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio
CHEBI:36946
NC(N)=S
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Suspected of damaging the unborn child.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep

GHS Signal Word: Warning

EINECSNumber : 200-543-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only