Learn More
Thiourea, 99%, extra pure, Thermo Scientific Chemicals
Thiourea, Thiocarbamide, CAS # 62-56-6, is an analogue of urea with a single sulfur-substituted oxygen.
$97.07 - $608.51
Chemical Identifiers
CAS | 62-56-6 |
---|---|
Molecular Formula | CH4N2S |
Molecular Weight (g/mol) | 76.12 |
MDL Number | MFCD00008067 |
InChI Key | UMGDCJDMYOKAJW-UHFFFAOYSA-N |
Synonym | thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio |
PubChem CID | 2723790 |
ChEBI | CHEBI:36946 |
IUPAC Name | thiourea |
SMILES | NC(N)=S |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC138915000
|
Thermo Scientific Chemicals
138915000 |
500 g | Plastic bottle |
Each for $97.07
|
|
||||
AC138910010
|
Thermo Scientific Chemicals
138910010 |
1 kg | Plastic bottle |
Each for $142.37
|
|
||||
AC138910050
|
Thermo Scientific Chemicals
138910050 |
5 kg | Plastic drum |
Each for $608.51
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• Thiourea is a derivative of the biological compound, urea
• Thiourea is highly toxic when it decomposes and acts as a carcinogen
Applications
• Thiourea can be used as a denaturant in lysis buffers and similar sample buffers that require solubilization of proteins
• Thiourea can act as an antioxidant and chromophore
• Thiourea can be used in the study of cellular redox, autophagy, and aging
Chemical Identifiers
62-56-6 | |
76.12 | |
UMGDCJDMYOKAJW-UHFFFAOYSA-N | |
2723790 | |
thiourea |
CH4N2S | |
MFCD00008067 | |
thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio | |
CHEBI:36946 | |
NC(N)=S |
Specifications
62-56-6 | |
White | |
Authentic | |
Plastic bottle | |
H2NCSNH2 | |
500 g | |
01,1164; 02,412; 03,290; 06,586; 11,519; 15,308 | |
thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio | |
UMGDCJDMYOKAJW-UHFFFAOYSA-N | |
thiourea | |
2723790 | |
76.12 | |
Extra Pure | |
Thiourea |
171.0°C to 175.0°C | |
0.5% max. | |
98.5% min. (Iodimetry) | |
CH4N2S | |
MFCD00008067 | |
03, 180 | |
15, 9521 | |
Solubility in water: 136g/L (20°C). Other solubilities: 12g/100g methanol (20°C),soluble in isopropylalcohol,butanol ,pentanol,sparingly soluble in ether | |
NC(N)=S | |
76.12 | |
CHEBI:36946 | |
99% | |
Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Suspected of damaging the unborn child.
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep
GHS Signal Word: Warning
EINECSNumber : 200-543-5
RUO – Research Use Only