missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thujone (alpha- and beta- mixture) 70.0+%, TCI America™
$121.11 - $356.68
Chemical Identifiers
CAS | 1125-12-8 |
---|---|
Molecular Formula | C10H16O |
Molecular Weight (g/mol) | 152.237 |
MDL Number | MFCD00135597 |
InChI Key | USMNOWBWPHYOEA-UHFFFAOYSA-N |
Synonym | 1-Isopropyl-4-methylbicyclo[3.1.0]hexan-3-one |
PubChem CID | 11027 |
ChEBI | CHEBI:50040 |
IUPAC Name | 4-methyl-1-propan-2-ylbicyclo[3.1.0]hexan-3-one |
SMILES | CC1C2CC2(CC1=O)C(C)C |
Chemical Identifiers
1125-12-8 | |
152.237 | |
USMNOWBWPHYOEA-UHFFFAOYSA-N | |
11027 | |
4-methyl-1-propan-2-ylbicyclo[3.1.0]hexan-3-one |
C10H16O | |
MFCD00135597 | |
1-Isopropyl-4-methylbicyclo[3.1.0]hexan-3-one | |
CHEBI:50040 | |
CC1C2CC2(CC1=O)C(C)C |
Specifications
1125-12-8 | |
C10H16O | |
5 mL | |
USMNOWBWPHYOEA-UHFFFAOYSA-N | |
4-methyl-1-propan-2-ylbicyclo[3.1.0]hexan-3-one | |
11027 | |
152.24 | |
Liquid |
Yellow | |
MFCD00135597 | |
1-Isopropyl-4-methylbicyclo[3.1.0]hexan-3-one | |
CC1C2CC2(CC1=O)C(C)C | |
152.237 | |
CHEBI:50040 | |
≥70.0% (GC) | |
Thujone (alpha- and beta- mixture) |
Safety and Handling
TSCA : Yes