missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thujone (alpha- and beta- mixture) 70.0+%, TCI America™
Supplier: TCI America T098925ML
Specifications
Thujone (alpha- and beta- mixture) | |
Yellow | |
MFCD00135597 | |
1-Isopropyl-4-methylbicyclo[3.1.0]hexan-3-one | |
CC1C2CC2(CC1=O)C(C)C | |
152.237 | |
CHEBI:50040 | |
≥70.0% (GC) |
1125-12-8 | |
C10H16O | |
25 mL | |
USMNOWBWPHYOEA-UHFFFAOYSA-N | |
4-methyl-1-propan-2-ylbicyclo[3.1.0]hexan-3-one | |
11027 | |
152.24 | |
Liquid |
Chemical Identifiers
1125-12-8 | |
152.237 | |
USMNOWBWPHYOEA-UHFFFAOYSA-N | |
11027 | |
4-methyl-1-propan-2-ylbicyclo[3.1.0]hexan-3-one |
C10H16O | |
MFCD00135597 | |
1-Isopropyl-4-methylbicyclo[3.1.0]hexan-3-one | |
CHEBI:50040 | |
CC1C2CC2(CC1=O)C(C)C |
Safety and Handling
TSCA : Yes