missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(II) chloride, 98%, anhydrous, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 196980250

 View more versions of this product

Catalog No. AC196980250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

652.0°C
246.0°C
7772-99-8
Fine Crystalline Powder or Flakes
97.5 to 102.5% (Iodimetry)
SnCl2
01,1113; 02,389; 05,631; 06,554; 11,521; 13,88; 15,138; 16,68; 17,334
Stannous chloride
AXZWODMDQAVCJE-UHFFFAOYSA-L
λ²-tin(2+) dichloride
24479
189.6
Glass bottle
3.9500g/mL
Tin(II) chloride
98%
White
25 g
Cl2Sn
MFCD00011241
15, 8910
Solubility in water: in water: 2700g/L (20°C). Other solubilities: soluble in alkalies, soluble in tartaric acid, alcohol, acetone, ether,, methyl acetate, methyl ethyl ketone and isobutyl, alcohol
[Cl-].[Cl-].[Sn++]
189.61
CHEBI:78067
98%
3.95

Chemical Identifiers

7772-99-8
189.61
AXZWODMDQAVCJE-UHFFFAOYSA-L
24479
[Cl-].[Cl-].[Sn++]
Cl2Sn
MFCD00011241
Stannous chloride
CHEBI:78067
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
May cause dam

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

GHS Signal Word: Danger

EINECSNumber : 231-868-0

RTECSNumber : XP8700000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only