Learn More
Tin(II) chloride, 98%, anhydrous, Thermo Scientific Chemicals
$62.64 - $356.23
Chemical Identifiers
CAS | 7772-99-8 |
---|---|
Molecular Formula | Cl2Sn |
Molecular Weight (g/mol) | 189.61 |
MDL Number | MFCD00011241 |
InChI Key | AXZWODMDQAVCJE-UHFFFAOYSA-L |
Synonym | Stannous chloride |
PubChem CID | 24479 |
ChEBI | CHEBI:78067 |
SMILES | [Cl-].[Cl-].[Sn++] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC196980250
|
Thermo Scientific Chemicals
196980250 |
25 g | Glass bottle |
Each for $62.64
|
|
||||
AC196981000
|
Thermo Scientific Chemicals
196981000 |
100 g | Glass bottle |
Each for $128.31
|
|
||||
AC196985000
|
Thermo Scientific Chemicals
196985000 |
500 g | Glass bottle |
Each for $356.23
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7772-99-8 | |
189.61 | |
AXZWODMDQAVCJE-UHFFFAOYSA-L | |
24479 | |
[Cl-].[Cl-].[Sn++] |
Cl2Sn | |
MFCD00011241 | |
Stannous chloride | |
CHEBI:78067 |
Specifications
652.0°C | |
7772-99-8 | |
Fine Crystalline Powder or Flakes | |
97.5 to 102.5% (Iodimetry) | |
SnCl2 | |
01,1113; 02,389; 05,631; 06,554; 11,521; 13,88; 15,138; 16,68; 17,334 | |
Stannous chloride | |
AXZWODMDQAVCJE-UHFFFAOYSA-L | |
λ²-tin(2+) dichloride | |
24479 | |
189.6 | |
Glass bottle | |
3.9500g/mL |
246.0°C | |
White | |
25 g | |
Cl2Sn | |
MFCD00011241 | |
15, 8910 | |
Solubility in water: in water: 2700g/L (20°C). Other solubilities: soluble in alkalies, soluble in tartaric acid, alcohol, acetone, ether,, methyl acetate, methyl ethyl ketone and isobutyl, alcohol | |
[Cl-].[Cl-].[Sn++] | |
189.61 | |
CHEBI:78067 | |
98% | |
3.95 | |
Tin(II) chloride, 98% |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
May cause dam
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
EINECSNumber : 231-868-0
RTECSNumber : XP8700000
TSCA : TSCA
RUO – Research Use Only