Learn More
Tin(II) chloride, 98%, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 196981000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
652.0°C | |
246.0°C | |
7772-99-8 | |
Fine Crystalline Powder or Flakes | |
97.5 to 102.5% (Iodimetry) | |
SnCl2 | |
01,1113; 02,389; 05,631; 06,554; 11,521; 13,88; 15,138; 16,68; 17,334 | |
Stannous chloride | |
AXZWODMDQAVCJE-UHFFFAOYSA-L | |
dichlorotin | |
24479 | |
189.6 | |
Glass bottle | |
3.9500g/mL |
Tin(II) chloride | |
98% | |
White | |
100 g | |
Cl2Sn | |
MFCD00011241 | |
15, 8910 | |
Solubility in water: in water: 2700g/L (20°C). Other solubilities: soluble in alkalies, soluble in tartaric acid, alcohol, acetone, ether,, methyl acetate, methyl ethyl ketone and isobutyl, alcohol | |
[Cl-].[Cl-].[Sn++] | |
189.61 | |
CHEBI:78067 | |
98% | |
3.95 |
Chemical Identifiers
7772-99-8 | |
189.61 | |
AXZWODMDQAVCJE-UHFFFAOYSA-L | |
24479 | |
[Cl-].[Cl-].[Sn++] |
Cl2Sn | |
MFCD00011241 | |
Stannous chloride | |
CHEBI:78067 |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
May cause dam
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
EINECSNumber : 231-868-0
RTECSNumber : XP8700000
TSCA : TSCA
RUO – Research Use Only