missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(II) chloride dihydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 424485000

 View more versions of this product

Catalog No. AC424485000



Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tin(II) chloride dihydrate
10025-69-1
500 g
Cl2Sn·2H2O
MFCD00149863
15, 8910
Solubility in water: 1187g/L (20°C). Other solubilities: soluble in alkalies and hcl, soluble in tartaric acid, alcohol, ethyl acetate, and glacial acetic acid
98.0 to 103.0%
Plastic bottle
White
Adhering Crystalline Powder or Crystals
98+%
SnCl2·2H2O
01,1113; 02,389; 05,631; 06,554; 11,480; 13,298; 15,309; 16,68
Stannous chloride dihydrate
225.63
ACS Reagent
2.7100g/mL

Chemical Identifiers

10025-69-1
Cl2Sn·2H2O
MFCD00149863
Stannous chloride dihydrate
Videos
Safety and Handling

Safety and Handling

GHS P Statement Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation. Suspected of causing genetic defects if swallowed. Suspected of damaging the unborn child. May cause damage to organs through prolonged or repeated exposure if swallowed. Very toxic to aquatic life.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Danger

RTECSNumber : XP8850000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only