missing translation for 'onlineSavingsMsg'
Learn More

Tin(II) chloride dihydrate, 98+%, ACS reagent

CAS: 10025-69-1 | Cl2Sn·2H2O | 225.63 g/mol

$96.13 - $472.44

Chemical Identifiers

CAS 10025-69-1
Molecular Formula Cl2Sn·2H2O
Molecular Weight (g/mol) 225.63
MDL Number MFCD00149863
Synonym Stannous chloride dihydrate
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC424480050
SDS View Documents
Thermo Scientific Chemicals
424480050
5 g Glass bottle
Each for $96.13
Only null left
 
AC424481000
SDS View Documents
Thermo Scientific Chemicals
424481000
100 g Plastic Bottle
Each for $213.05
Only null left
 
AC424485000
SDS View Documents
Thermo Scientific Chemicals
424485000
500 g Plastic bottle
Each for $472.44
Case of 6 Each for $0.00
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10025-69-1
225.63
Stannous chloride dihydrate
Cl2Sn·2H2O
MFCD00149863

Specifications

652.0°C
10025-69-1
Adhering Crystalline Powder or Crystals
98.0 to 103% (Iodimetry)
SnCl2·2H2O
01,1113; 02,389; 05,631; 06,554; 11,480; 13,298; 15,309; 16,68
Stannous chloride dihydrate
225.63
≥98%
Glass bottle
Tin(II) chloride dihydrate
37.0°C to 38.0°C
White
5 g
Cl2Sn·2H2O
MFCD00149863
15, 8910
(in hydrochloric acid) Passes test
225.63
ACS Reagent
2.7100g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Harmful if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
May cause damage to organs through prolon

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

GHS Signal Word: Danger

RTECSNumber : XP8850000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only