Learn More
Tin(II) fluoride, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 308501000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tin(II) fluoride | |
7783-47-3 | |
100 g | |
F2Sn | |
MFCD00042540 | |
Solubility in water: 300- 390g/L. Other solubilities: insoluble in ethanol,diethyl ether,chloroform | |
156.69 | |
Plastic bottle |
White | |
Powder, Crystals or Crystalline Powder | |
99% | |
SnF2 | |
15, 8911 | |
156.69 | |
99% |
Chemical Identifiers
7783-47-3 | |
F2Sn | |
156.69 | |
MFCD00042540 |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Danger
EINECSNumber : 231-999-3
RUO – Research Use Only