missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(II) fluoride, 99%, Thermo Scientific Chemicals

$58.82 - $483.46

Chemical Identifiers

CAS 7783-47-3
Molecular Formula F2Sn
Molecular Weight (g/mol) 156.69
MDL Number MFCD00042540
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC308500050
SDS View Documents
Thermo Scientific Chemicals
308500050
5 g Glass bottle N/A
 
AC308501000
SDS View Documents
Thermo Scientific Chemicals
308501000
100 g Plastic bottle N/A
 
AC308505000
SDS View Documents
Thermo Scientific Chemicals
308505000
500 g Plastic bottle N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7783-47-3
F2Sn
156.69
MFCD00042540

Specifications

White
Powder, Crystals or Crystalline Powder
99%
SnF2
15, 8911
156.69
99%
Tin(II) fluoride
7783-47-3
5 g
F2Sn
MFCD00042540
Solubility in water: 300- 390g/L. Other solubilities: insoluble in ethanol,diethyl ether,chloroform
156.69
Glass bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Danger

EINECSNumber : 231-999-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only