missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(II) fluoride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 308505000

 View more versions of this product

Catalog No. AC308505000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tin(II) fluoride
7783-47-3
500 g
F2Sn
MFCD00042540
Solubility in water: 300- 390g/L. Other solubilities: insoluble in ethanol,diethyl ether,chloroform
156.69
Plastic bottle
White
Powder, Crystals or Crystalline Powder
99%
SnF2
15, 8911
156.69
99%

Chemical Identifiers

7783-47-3
F2Sn
156.69
MFCD00042540
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Danger

EINECSNumber : 231-999-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only