Learn More
Tin(II) sulfate, 95%
CAS: 7488-55-3 | O4SSn | 214.77 g/mol
$89.36 - $281.91
Chemical Identifiers
| CAS | 7488-55-3 |
|---|---|
| Molecular Formula | O4SSn |
| Molecular Weight (g/mol) | 214.77 |
| MDL Number | MFCD00011246 |
| InChI Key | OBBXFSIWZVFYJR-UHFFFAOYSA-L |
| Synonym | stannous sulfate, tin ii sulfate, tin 2+ sulfate, unii-0mfe10j96e, sulfuric acid, tin 2+ salt 1:1, stannous sulfate, crystal, stannous sulfate, solutions, tin, ion sn2+ sulfate, tin ii sulfate 250g |
| PubChem CID | 62643 |
| SMILES | [Sn++].[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC223661000
|
Thermo Scientific Chemicals
223661000 |
100 g | Glass bottle |
Each for $89.36
|
|
||||
|
AC223665000
|
Thermo Scientific Chemicals
223665000 |
500 g | Glass bottle |
Each for $281.91
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7488-55-3 | |
| 214.77 | |
| OBBXFSIWZVFYJR-UHFFFAOYSA-L | |
| 62643 |
| O4SSn | |
| MFCD00011246 | |
| stannous sulfate, tin ii sulfate, tin 2+ sulfate, unii-0mfe10j96e, sulfuric acid, tin 2+ salt 1:1, stannous sulfate, crystal, stannous sulfate, solutions, tin, ion sn2+ sulfate, tin ii sulfate 250g | |
| [Sn++].[O-]S([O-])(=O)=O |
Specifications
| 360.0°C | |
| White | |
| 100 g | |
| O4SSn | |
| MFCD00011246 | |
| stannous sulfate, tin ii sulfate, tin 2+ sulfate, unii-0mfe10j96e, sulfuric acid, tin 2+ salt 1:1, stannous sulfate, crystal, stannous sulfate, solutions, tin, ion sn2+ sulfate, tin ii sulfate 250g | |
| OBBXFSIWZVFYJR-UHFFFAOYSA-L | |
| tin(2+);sulfate | |
| 62643 | |
| 95% | |
| Tin(II) sulfate |
| 7488-55-3 | |
| Powder, Crystals and/or Lumps | |
| 94% min. (Iodimetry) | |
| SnSO4 | |
| 15,8917 | |
| Solubility in water: soluble. Other solubilities: soluble in diluted sulfuric acid | |
| [Sn++].[O-]S([O-])(=O)=O | |
| 214.77 | |
| 214.75 | |
| Glass bottle |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful if inhaled.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated ex
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minu
GHS Signal Word: Danger
EINECSNumber : 231-302-2
RUO – Research Use Only