missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tin(IV) oxide, 15% in H{2}O colloidal dispersion, Thermo Scientific Chemicals

Tin(IV) oxide, O2Sn, CAS Number-18282-10-5, white tin oxide, tin iv oxide, tin peroxide, stannic dioxide, stannic oxide, tin oxide sno2, tin dioxide, stannic anhydride, cassiterite, flowers of tin, 500g, 15% in H2O colloidal dispersion, CHEBI:52991, 1.153 g/mL, 150.69, XOLBLPGZBRYERU-UHFFFAOYSA-N

Supplier:  Thermo Scientific Chemicals 044592A3

 View more versions of this product

Catalog No. AA44592A3


Explore available promotions

Description

Description

It is used to detect carbon monoxide. Used in ceramic glazes and colors, putty, perfume preparations, textiles, polishing powder for steel, and the manufacture of special glasses. Stannic oxide is also used in making white patinas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tin(IV) oxide
18282-10-5
2 kg
O2Sn
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
XOLBLPGZBRYERU-UHFFFAOYSA-N
stannanedione
29011
150.69
1.153
colloidal dispersion
Liquid
15% in H2O
MFCD00011244
Fully miscible in water.
O=[Sn]=O
150.71
CHEBI:52991
Odorless

Chemical Identifiers

18282-10-5
150.71
XOLBLPGZBRYERU-UHFFFAOYSA-N
29011
stannanedione
O2Sn
MFCD00011244
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
CHEBI:52991
O=[Sn]=O
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only