missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tin(IV) oxide, 15% in H{2}O colloidal dispersion, Thermo Scientific Chemicals
Tin(IV) oxide, O2Sn, CAS Number-18282-10-5, white tin oxide, tin iv oxide, tin peroxide, stannic dioxide, stannic oxide, tin oxide sno2, tin dioxide, stannic anhydride, cassiterite, flowers of tin, 500g, 15% in H2O colloidal dispersion, CHEBI:52991, 1.153 g/mL, 150.69, XOLBLPGZBRYERU-UHFFFAOYSA-N
$473.08 - $473.08
Chemical Identifiers
CAS | 18282-10-5 |
---|---|
Molecular Formula | O2Sn |
Molecular Weight (g/mol) | 150.71 |
MDL Number | MFCD00011244 |
InChI Key | XOLBLPGZBRYERU-UHFFFAOYSA-N |
Synonym | tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride |
PubChem CID | 29011 |
ChEBI | CHEBI:52991 |
IUPAC Name | stannanedione |
SMILES | O=[Sn]=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA44592A3
|
Thermo Scientific Chemicals
044592A3 |
2 kg |
Each for $473.08
|
|
|||||
Description
It is used to detect carbon monoxide. Used in ceramic glazes and colors, putty, perfume preparations, textiles, polishing powder for steel, and the manufacture of special glasses. Stannic oxide is also used in making white patinas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18282-10-5 | |
150.71 | |
XOLBLPGZBRYERU-UHFFFAOYSA-N | |
29011 | |
stannanedione |
O2Sn | |
MFCD00011244 | |
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride | |
CHEBI:52991 | |
O=[Sn]=O |
Specifications
18282-10-5 | |
2 kg | |
O2Sn | |
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride | |
XOLBLPGZBRYERU-UHFFFAOYSA-N | |
stannanedione | |
29011 | |
150.69 | |
1.153 |
Liquid | |
15% in H2O | |
MFCD00011244 | |
Fully miscible in water. | |
O=[Sn]=O | |
150.71 | |
CHEBI:52991 | |
Odorless | |
Tin(IV) oxide, colloidal dispersion |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only