missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tinidazole 98.0+%, TCI America™
$81.32 - $190.32
Chemical Identifiers
CAS | 19387-91-8 |
---|---|
Molecular Formula | C8H13N3O4S |
Molecular Weight (g/mol) | 247.27 |
MDL Number | MFCD00057217 |
InChI Key | HJLSLZFTEKNLFI-UHFFFAOYSA-N |
Synonym | 1-[2-(Ethylsulfonyl)ethyl]-2-methyl-5-nitroimidazole |
PubChem CID | 5479 |
ChEBI | CHEBI:63627 |
IUPAC Name | 1-[2-(ethanesulfonyl)ethyl]-2-methyl-5-nitro-1H-imidazole |
SMILES | CCS(=O)(=O)CCN1C(C)=NC=C1[N+]([O-])=O |
Chemical Identifiers
19387-91-8 | |
247.27 | |
HJLSLZFTEKNLFI-UHFFFAOYSA-N | |
5479 | |
1-[2-(ethanesulfonyl)ethyl]-2-methyl-5-nitro-1H-imidazole |
C8H13N3O4S | |
MFCD00057217 | |
1-[2-(Ethylsulfonyl)ethyl]-2-methyl-5-nitroimidazole | |
CHEBI:63627 | |
CCS(=O)(=O)CCN1C(C)=NC=C1[N+]([O-])=O |
Specifications
19387-91-8 | |
White-Yellow | |
MFCD00057217 | |
1-[2-(Ethylsulfonyl)ethyl]-2-methyl-5-nitroimidazole | |
CCS(=O)(=O)CCN1C(C)=NC=C1[N+]([O-])=O | |
247.27 | |
CHEBI:63627 | |
≥98.0% (HPLC,T) | |
Tinidazole |
129°C | |
C8H13N3O4S | |
5 g | |
HJLSLZFTEKNLFI-UHFFFAOYSA-N | |
1-[2-(ethanesulfonyl)ethyl]-2-methyl-5-nitro-1H-imidazole | |
5479 | |
247.27 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : NI6255000
TSCA : No