missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tinidazole 98.0+%, TCI America™

Supplier:  TCI America T305825G

 View more versions of this product

Catalog No. T305825G


Explore available promotions

Description

Description

Specifications

Specifications

Tinidazole
129°C
C8H13N3O4S
25 g
HJLSLZFTEKNLFI-UHFFFAOYSA-N
1-[2-(ethanesulfonyl)ethyl]-2-methyl-5-nitro-1H-imidazole
5479
247.27
Crystalline Powder
19387-91-8
White-Yellow
MFCD00057217
1-[2-(Ethylsulfonyl)ethyl]-2-methyl-5-nitroimidazole
CCS(=O)(=O)CCN1C(C)=NC=C1[N+]([O-])=O
247.27
CHEBI:63627
≥98.0% (HPLC,T)

Chemical Identifiers

19387-91-8
247.27
HJLSLZFTEKNLFI-UHFFFAOYSA-N
5479
1-[2-(ethanesulfonyl)ethyl]-2-methyl-5-nitro-1H-imidazole
C8H13N3O4S
MFCD00057217
1-[2-(Ethylsulfonyl)ethyl]-2-methyl-5-nitroimidazole
CHEBI:63627
CCS(=O)(=O)CCN1C(C)=NC=C1[N+]([O-])=O
Videos
Safety and Handling

Safety and Handling

RTECSNumber : NI6255000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions