missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(III) chloride tetrahydrofuran complex, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 375160100

 View more versions of this product

Catalog No. AC375160100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Specifications

Titanium(III) chloride tetrahydrofuran complex
Blue to Blue-Green
Crystals or Powder
96% min. (Argentometry)
MFCD01631276
Solubility in water: reacts violently
C1CCOC1.C1CCOC1.C1CCOC1.Cl[Ti](Cl)Cl
370.56
370.56
Authentic
>250°C
18039-90-2
10 g
C12H24Cl3O3Ti
oxolane; trichlorotitanium, titanium iii chloride tetrahydrofuran complex 1:3, trichlorotris thf titanium, titanium trichloride tri thf, trichlorotris tetrahydrofuran titanium, thf-titanium trichloride complex, 3:1, titanium trichloride tris tetrahydrofuran, trichlorotris tetrahydrofuran titanium iii, trichlorotris tetrahydrofuran tita-nium iii, titanium trichloride tetrahydrofuran complex 1:3
MIDYWIPTCYRMQF-UHFFFAOYSA-K
oxolane;trichlorotitanium
6097232
97%
Glass Bottle

Chemical Identifiers

18039-90-2
370.56
MIDYWIPTCYRMQF-UHFFFAOYSA-K
6097232
C1CCOC1.C1CCOC1.C1CCOC1.Cl[Ti](Cl)Cl
C12H24Cl3O3Ti
MFCD01631276
oxolane; trichlorotitanium, titanium iii chloride tetrahydrofuran complex 1:3, trichlorotris thf titanium, titanium trichloride tri thf, trichlorotris tetrahydrofuran titanium, thf-titanium trichloride complex, 3:1, titanium trichloride tris tetrahydrofuran, trichlorotris tetrahydrofuran titanium iii, trichlorotris tetrahydrofuran tita-nium iii, titanium trichloride tetrahydrofuran complex 1:3
oxolane;trichlorotitanium
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable solid.
Causes serious eye damage.
Reacts violently with water.
May form explosive peroxides.
Contact with water liberates toxic gas.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only