Learn More
Titanium(III) chloride tetrahydrofuran complex, 97%
CAS: 18039-90-2 | C12H24Cl3O3Ti | 370.56 g/mol
$103.63 - $366.01
Chemical Identifiers
| CAS | 18039-90-2 |
|---|---|
| Molecular Formula | C12H24Cl3O3Ti |
| Molecular Weight (g/mol) | 370.56 |
| MDL Number | MFCD01631276 |
| InChI Key | MIDYWIPTCYRMQF-UHFFFAOYSA-K |
| Synonym | oxolane; trichlorotitanium, titanium iii chloride tetrahydrofuran complex 1:3, trichlorotris thf titanium, titanium trichloride tri thf, trichlorotris tetrahydrofuran titanium, thf-titanium trichloride complex, 3:1, titanium trichloride tris tetrahydrofuran, trichlorotris tetrahydrofuran titanium iii, trichlorotris tetrahydrofuran tita-nium iii, titanium trichloride tetrahydrofuran complex 1:3 |
| PubChem CID | 6097232 |
| IUPAC Name | oxolane;trichlorotitanium |
| SMILES | C1CCOC1.C1CCOC1.C1CCOC1.Cl[Ti](Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC375160010
|
Thermo Scientific Chemicals
375160010 |
1 g | Glass bottle |
Each for $103.63
|
|
||||
|
AC375160100
|
Thermo Scientific Chemicals
375160100 |
10 g | Glass Bottle |
Each for $366.01
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
| 18039-90-2 | |
| 370.56 | |
| MIDYWIPTCYRMQF-UHFFFAOYSA-K | |
| 6097232 | |
| C1CCOC1.C1CCOC1.C1CCOC1.Cl[Ti](Cl)Cl |
| C12H24Cl3O3Ti | |
| MFCD01631276 | |
| oxolane; trichlorotitanium, titanium iii chloride tetrahydrofuran complex 1:3, trichlorotris thf titanium, titanium trichloride tri thf, trichlorotris tetrahydrofuran titanium, thf-titanium trichloride complex, 3:1, titanium trichloride tris tetrahydrofuran, trichlorotris tetrahydrofuran titanium iii, trichlorotris tetrahydrofuran tita-nium iii, titanium trichloride tetrahydrofuran complex 1:3 | |
| oxolane;trichlorotitanium |
Specifications
| >250°C | |
| 18039-90-2 | |
| 1 g | |
| C12H24Cl3O3Ti | |
| oxolane; trichlorotitanium, titanium iii chloride tetrahydrofuran complex 1:3, trichlorotris thf titanium, titanium trichloride tri thf, trichlorotris tetrahydrofuran titanium, thf-titanium trichloride complex, 3:1, titanium trichloride tris tetrahydrofuran, trichlorotris tetrahydrofuran titanium iii, trichlorotris tetrahydrofuran tita-nium iii, titanium trichloride tetrahydrofuran complex 1:3 | |
| MIDYWIPTCYRMQF-UHFFFAOYSA-K | |
| oxolane;trichlorotitanium | |
| 6097232 | |
| 97% | |
| Glass bottle |
| Blue to Blue-Green | |
| Crystals or Powder | |
| 96% min. (Argentometry) | |
| MFCD01631276 | |
| Solubility in water: reacts violently | |
| C1CCOC1.C1CCOC1.C1CCOC1.Cl[Ti](Cl)Cl | |
| 370.56 | |
| 370.56 | |
| Authentic | |
| Titanium(III) chloride tetrahydrofuran complex |
Safety and Handling
GHS H Statement:
Flammable solid.
Causes serious eye damage.
Reacts violently with water.
May form explosive peroxides.
Contact with water liberates toxic gas.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only