Learn More
Titanium(IV) chloride, 99.9%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 197231000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
136.5°C | |
-24.0°C | |
Yellow | |
100 mL | |
Cl4Ti | |
MFCD00011267 | |
15, 9634 | |
Solubility in water: reacts forming hcl. Other solubilities: miscible with cold cond. hcl, soluble in alcohol (reacts, forming hcl), miscible with aliphatic hydrocarbons and, chlorinated aliphatic hydrocarbons | |
[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4] | |
189.71 | |
189.71 | |
Glass bottle | |
1.7200g/mL |
Titanium(IV) chloride | |
7550-45-0 | |
Liquid | |
99.85% min | |
TiCl4 | |
01,1169; 02,414; 03,291; 04,507; 05,671; 06,590; 07,370; 08,483; 09,468; 10,401; 11,337; 12,94; 13,6; 14,111; 15,317; 16,332; 17,344 | |
titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4 | |
XJDNKRIXUMDJCW-UHFFFAOYSA-J | |
titanium(4+);tetrachloride | |
160960 | |
99.9% | |
1.72 |
Chemical Identifiers
7550-45-0 | |
XJDNKRIXUMDJCW-UHFFFAOYSA-J | |
160960 | |
[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4] |
MFCD00011267 | |
titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4 | |
titanium(4+);tetrachloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
May cause respiratory irritation.
Reacts violently with water.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
EINECSNumber : 231-441-9
RTECSNumber : XR1925000
TSCA : TSCA
Recommended Storage : note: product may darken on exposure to light
RUO – Research Use Only