missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 223760500

 View more versions of this product

Catalog No. AC223760500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Specifications

Titanium(IV) ethoxide
-40.0°C
1.0900g/mL
33°C
Glass bottle
1.5050 to 1.5070
50 g
1.09
Solubility in water: hydrolysis.
CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]
228.15
60-80 mPa.s (20°C)
33-35% TiO2
3087-36-3
Colorless to Yellow
150.0°C to 152.0°C (10.0 mmHg)
Authentic
C8H20O4Ti
Ti(OC2H5)4
01,III,1338
titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
JMXKSZRRTHPKDL-UHFFFAOYSA-N
ethanolate;titanium(4+)
76524
228.15
Liquid

Chemical Identifiers

3087-36-3
228.15
titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
ethanolate;titanium(4+)
C8H20O4Ti
JMXKSZRRTHPKDL-UHFFFAOYSA-N
76524
CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear eye protection/face protection.
Wash f

GHS Signal Word: Warning

EINECSNumber : 221-410-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only