missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific Chemicals

$111.03 - $309.19

Chemical Identifiers

CAS 3087-36-3
Molecular Formula C8H20O4Ti
Molecular Weight (g/mol) 228.15
InChI Key JMXKSZRRTHPKDL-UHFFFAOYSA-N
Synonym titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
PubChem CID 76524
IUPAC Name ethanolate;titanium(4+)
SMILES CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC223760500
SDS View Documents
Thermo Scientific Chemicals
223760500
50 g Glass bottle
Each for $111.03
 
AC223762500
SDS View Documents
Thermo Scientific Chemicals
223762500
250 g Glass bottle
Each for $309.19
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Chemical Identifiers

3087-36-3
228.15
titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
ethanolate;titanium(4+)
C8H20O4Ti
JMXKSZRRTHPKDL-UHFFFAOYSA-N
76524
CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]

Specifications

3087-36-3
Colorless to Yellow
150.0°C to 152.0°C (10.0 mmHg)
Authentic
C8H20O4Ti
Ti(OC2H5)4
01,III,1338
titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
JMXKSZRRTHPKDL-UHFFFAOYSA-N
ethanolate;titanium(4+)
76524
228.15
Liquid
-40.0°C
1.0900g/mL
33°C
Glass bottle
1.5050 to 1.5070
50 g
1.09
Solubility in water: hydrolysis.
CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]
228.15
60-80 mPa.s (20°C)
33-35% TiO2
Titanium(IV) ethoxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear eye protection/face protection.
Wash f

GHS Signal Word: Warning

EINECSNumber : 221-410-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only